Product Details

SNP ID
rs140909896
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:73987524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAAACAGCAACTTACATCACTT[C/T]GACTTGGTTGCAATGGGTTCCTTTC
Phenotype
MIM: 600324 MIM: 173460 MIM: 121010
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CXCL5 PubMed Links

Gene Details

Gene
CXCL5
Gene Name
C-X-C motif chemokine ligand 5
There are no transcripts associated with this gene.

Gene
PF4
Gene Name
platelet factor 4
There are no transcripts associated with this gene.

Gene
PPBP
Gene Name
pro-platelet basic protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002704.3 364 Missense Mutation AAA,GAA K93E NP_002695.1

View Full Product Details