Product Details
- SNP ID
-
rs141265672
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:53459425 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGAGCCTGCCCCTGAACAGGAGAG[C/G]ACCGAAGCTACACCTGCAGAATAGG
- Phenotype
-
MIM: 607686
MIM: 609732
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FIP1L1
PubMed Links
Gene Details
- Gene
- FIP1L1
- Gene Name
- factor interacting with PAPOLA and CPSF1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001134937.1 |
4173 |
Missense Mutation |
AGC,AGG |
S581R |
NP_001128409.1 |
NM_001134938.1 |
4173 |
Missense Mutation |
AGC,AGG |
S513R |
NP_001128410.1 |
NM_030917.3 |
4173 |
Missense Mutation |
AGC,AGG |
S587R |
NP_112179.2 |
XM_005265769.4 |
4173 |
Missense Mutation |
AGC,AGG |
S596R |
XP_005265826.1 |
XM_005265773.4 |
4173 |
Missense Mutation |
AGC,AGG |
S573R |
XP_005265830.1 |
XM_005265774.4 |
4173 |
Missense Mutation |
AGC,AGG |
S564R |
XP_005265831.1 |
XM_005265778.4 |
4173 |
Missense Mutation |
GCA,GGA |
A525G |
XP_005265835.1 |
XM_005265779.4 |
4173 |
Missense Mutation |
GCA,GGA |
A516G |
XP_005265836.1 |
XM_005265781.4 |
4173 |
Missense Mutation |
AGC,AGG |
S537R |
XP_005265838.1 |
XM_005265782.4 |
4173 |
Missense Mutation |
AGC,AGG |
S528R |
XP_005265839.1 |
XM_017008662.1 |
4173 |
Missense Mutation |
AGC,AGG |
S572R |
XP_016864151.1 |
XM_017008663.1 |
4173 |
Missense Mutation |
AGC,AGG |
S560R |
XP_016864152.1 |
XM_017008664.1 |
4173 |
Missense Mutation |
AGC,AGG |
S558R |
XP_016864153.1 |
XM_017008665.1 |
4173 |
Missense Mutation |
AGC,AGG |
S551R |
XP_016864154.1 |
XM_017008666.1 |
4173 |
Missense Mutation |
AGC,AGG |
S549R |
XP_016864155.1 |
XM_017008667.1 |
4173 |
Missense Mutation |
AGC,AGG |
S545R |
XP_016864156.1 |
XM_017008668.1 |
4173 |
Missense Mutation |
AGC,AGG |
S536R |
XP_016864157.1 |
XM_017008669.1 |
4173 |
Missense Mutation |
AGC,AGG |
S522R |
XP_016864158.1 |
XM_017008670.1 |
4173 |
Missense Mutation |
GCA,GGA |
A493G |
XP_016864159.1 |
XM_017008671.1 |
4173 |
Missense Mutation |
GCA,GGA |
A466G |
XP_016864160.1 |
- Gene
- LNX1
- Gene Name
- ligand of numb-protein X 1
View Full Product Details