Product Details

SNP ID
rs141746561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:53462822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATGAATTTCATTAAATTTCCTA[C/T]TGTAAAATATTTATCTGCTGCATTT
Phenotype
MIM: 607686 MIM: 609732
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FIP1L1 PubMed Links

Gene Details

Gene
FIP1L1
Gene Name
factor interacting with PAPOLA and CPSF1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134937.1 Intron NP_001128409.1
NM_001134938.1 Intron NP_001128410.1
NM_030917.3 Intron NP_112179.2
XM_005265769.4 Intron XP_005265826.1
XM_005265773.4 Intron XP_005265830.1
XM_005265774.4 Intron XP_005265831.1
XM_005265778.4 Intron XP_005265835.1
XM_005265779.4 Intron XP_005265836.1
XM_005265781.4 Intron XP_005265838.1
XM_005265782.4 Intron XP_005265839.1
XM_017008662.1 Intron XP_016864151.1
XM_017008663.1 Intron XP_016864152.1
XM_017008664.1 Intron XP_016864153.1
XM_017008665.1 Intron XP_016864154.1
XM_017008666.1 Intron XP_016864155.1
XM_017008667.1 Intron XP_016864156.1
XM_017008668.1 Intron XP_016864157.1
XM_017008669.1 Intron XP_016864158.1
XM_017008670.1 Intron XP_016864159.1
XM_017008671.1 Intron XP_016864160.1
Gene
LNX1
Gene Name
ligand of numb-protein X 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126328.2 Intron NP_001119800.1
NM_032622.2 Intron NP_116011.2
XM_005265785.4 Intron XP_005265842.1
XM_017008776.1 Intron XP_016864265.1

View Full Product Details