Product Details

SNP ID
rs142101131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:4268580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCATTGTCTGGGGCCTGTTTCA[C/G]AATTGCTTTAATAGTTCCCTTCCAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LYAR PubMed Links

Gene Details

Gene
LYAR
Gene Name
Ly1 antibody reactive
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145725.1 1129 Missense Mutation CTG,GTG L319V NP_001139197.1
NM_017816.2 1129 Missense Mutation CTG,GTG L319V NP_060286.1
XM_011513505.1 1129 Missense Mutation CTG,GTG L319V XP_011511807.1
XM_011513506.2 1129 Missense Mutation CTG,GTG L319V XP_011511808.1

View Full Product Details