Product Details

SNP ID
rs142214180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70993896 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGCCAGCTCTGAGGGGACCCGC[A/G]TCAAGAAAATCTCCATCGAAGGGAA
Phenotype
MIM: 125450 MIM: 609282
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCK PubMed Links

Gene Details

Gene
DCK
Gene Name
deoxycytidine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000788.2 349 Missense Mutation ATC,GTC I21V NP_000779.1
Gene
MOB1B
Gene Name
MOB kinase activator 1B
There are no transcripts associated with this gene.

View Full Product Details