Product Details

SNP ID
rs142766178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:2236308 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCACATCCTTATTCCGTTTGGACA[A/G]AAAGAAAGAATGTTCTTGAGCAGAT
Phenotype
MIM: 613430 MIM: 610887
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HAUS3 PubMed Links

Gene Details

Gene
HAUS3
Gene Name
HAUS augmin like complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303143.1 2030 Silent Mutation CTG,TTG L500L NP_001290072.1
NM_024511.6 2030 Silent Mutation CTG,TTG L500L NP_078787.2
Gene
POLN
Gene Name
DNA polymerase nu
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181808.3 2030 Intron NP_861524.2

View Full Product Details