Product Details

SNP ID
rs142927427
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:2072222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGACTCAGTGCGACATGGGCCTGG[C/T]GGAGGGCCCCAGGCCTCCTGCAGTG
Phenotype
MIM: 610647 MIM: 610887
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NAT8L PubMed Links

Gene Details

Gene
NAT8L
Gene Name
N-acetyltransferase 8 like
There are no transcripts associated with this gene.

Gene
POLN
Gene Name
DNA polymerase nu
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181808.3 2992 Silent Mutation CCA,CCG P865P NP_861524.2

View Full Product Details