Product Details

SNP ID
rs143698630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99948818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTAGAAGTTAACATACCACCACC[A/G]GCAATTGTAGCCTTGATGAGAGAAT
Phenotype
MIM: 142763
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJB14 PubMed Links

Gene Details

Gene
DNAJB14
Gene Name
DnaJ heat shock protein family (Hsp40) member B14
There are no transcripts associated with this gene.

Gene
H2AFZ
Gene Name
H2A histone family member Z
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002106.3 Intron NP_002097.1
Gene
LOC256880
Gene Name
uncharacterized LOC256880
There are no transcripts associated with this gene.

View Full Product Details