Product Details

SNP ID
rs148513455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:10442049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGAAGAAACAGTGGGCAAGGAA[A/G]TTGTGCTAGTCCACAGGAGGGCCGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF518B PubMed Links

Gene Details

Gene
ZNF518B
Gene Name
zinc finger protein 518B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053042.2 7225 UTR 3 NP_444270.2
XM_005248193.2 7225 UTR 3 XP_005248250.1
XM_017008784.1 7225 UTR 3 XP_016864273.1
XM_017008785.1 7225 UTR 3 XP_016864274.1
XM_017008786.1 7225 UTR 3 XP_016864275.1

View Full Product Details