Product Details

SNP ID
rs150542973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:11399178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTATTGAGTAGTTTGGGATCGAC[C/T]TGGGGGTGCGCCCGGCCTTTGGACT
Phenotype
MIM: 603244
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HS3ST1 PubMed Links

Gene Details

Gene
HS3ST1
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005114.2 5910 Silent Mutation CAA,CAG Q276Q NP_005105.1
XM_011513913.2 5910 Silent Mutation CAA,CAG Q276Q XP_011512215.1

View Full Product Details