Product Details

SNP ID
rs138513605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132814961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGATCCAGGCCTGCGAGGCACCA[C/T]GGAGCCAGATGCGACCGGTGGTGGA
Phenotype
MIM: 611179
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SHROOM1 PubMed Links

Gene Details

Gene
SHROOM1
Gene Name
shroom family member 1
There are no transcripts associated with this gene.

Gene
SOWAHA
Gene Name
sosondowah ankyrin repeat domain family member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175873.5 1638 Missense Mutation ACG,ATG T447M NP_787069.4

View Full Product Details