Product Details

SNP ID
rs139350220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:24491600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGTAGAATGATGATGCAGAGGA[C/G]GATGGCGATCAAGGCCCCAGTGCTG
Phenotype
MIM: 604555
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDH10 PubMed Links

Gene Details

Gene
CDH10
Gene Name
cadherin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317222.1 1861 Missense Mutation CTC,GTC L24V NP_001304151.1
NM_001317224.1 1861 Missense Mutation CTC,GTC L618V NP_001304153.1
NM_006727.4 1861 Missense Mutation CTC,GTC L618V NP_006718.2
XM_011513923.2 1861 Missense Mutation CTC,GTC L618V XP_011512225.1
XM_017008915.1 1861 Missense Mutation CTC,GTC L618V XP_016864404.1

View Full Product Details