Product Details

SNP ID
rs139894073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:72444170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGCTGCCCTGGGCACTCTCCCC[A/G]TCTGACTGGAACACCGGCACCTTGG
Phenotype
MIM: 610159
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF366 PubMed Links

Gene Details

Gene
ZNF366
Gene Name
zinc finger protein 366
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152625.2 2007 Silent Mutation GAC,GAT D607D NP_689838.1

View Full Product Details