Product Details

SNP ID
rs140667298
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:58494643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGTGTTTGGCACTGGAAACACC[A/G]TGTTGCCACACGATTTACCTTACCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GAPT PubMed Links

Gene Details

Gene
GAPT
Gene Name
GRB2-binding adaptor protein, transmembrane
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304428.1 632 Missense Mutation CAT,CGT H36R NP_001291357.1
NM_001304429.1 632 Missense Mutation CAT,CGT H36R NP_001291358.1
NM_001304431.1 632 Missense Mutation CAT,CGT H36R NP_001291360.1
NM_152687.3 632 Missense Mutation CAT,CGT H36R NP_689900.1

View Full Product Details