Product Details

SNP ID
rs141258579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140835475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCCCTATTCCAGAGGACGCCCA[A/T]CCAGGTACCGTCATCACATTGATTA
Phenotype
MIM: 606307 MIM: 606308 MIM: 606309 MIM: 606310 MIM: 606311 MIM: 606312 MIM: 606313 MIM: 606314
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PCDHA1 PubMed Links

Gene Details

Gene
PCDHA1
Gene Name
protocadherin alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018900.3 1092 Intron NP_061723.1
NM_031410.2 1092 Intron NP_113598.1
NM_031411.2 1092 Intron NP_113599.1
Gene
PCDHA2
Gene Name
protocadherin alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018905.2 1092 Intron NP_061728.1
NM_031495.1 1092 Intron NP_113683.1
NM_031496.1 1092 Intron NP_113684.1
Gene
PCDHA3
Gene Name
protocadherin alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018906.2 1092 Intron NP_061729.1
NM_031497.1 1092 Intron NP_113685.1
Gene
PCDHA4
Gene Name
protocadherin alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018907.3 1092 Intron NP_061730.1
NM_031500.2 1092 Intron NP_113688.1
Gene
PCDHA5
Gene Name
protocadherin alpha 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018908.2 1092 Intron NP_061731.1
NM_031501.1 1092 Intron NP_113689.1
Gene
PCDHA6
Gene Name
protocadherin alpha 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018909.3 1092 Intron NP_061732.1
NM_031848.2 1092 Intron NP_114036.1
NM_031849.2 1092 Intron NP_114037.1
Gene
PCDHA7
Gene Name
protocadherin alpha 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018910.2 1092 Silent Mutation CAA,CAT Q364H NP_061733.1
NM_031852.1 1092 Silent Mutation CAA,CAT Q364H NP_114040.1
Gene
PCDHA8
Gene Name
protocadherin alpha 8
There are no transcripts associated with this gene.

View Full Product Details