Product Details

SNP ID
rs141294003
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:111072911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCAAAAGTACCGAGTTCAACAA[A/C]ACCGTCTCTTGTAGCAATCGGGTGA
Phenotype
MIM: 607003
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TSLP PubMed Links

Gene Details

Gene
TSLP
Gene Name
thymic stromal lymphopoietin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033035.4 394 Missense Mutation AAA,AAC K65N NP_149024.1
NM_138551.4 394 Intron NP_612561.2
XM_011543698.1 394 Intron XP_011542000.1

View Full Product Details