Product Details

SNP ID
rs141800763
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:111071794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCAATGAGAGGCAAAACCTGGT[C/G]CTTGAGCACTGGCCCCTAAGGCAGG
Phenotype
MIM: 607003
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TSLP PubMed Links

Gene Details

Gene
TSLP
Gene Name
thymic stromal lymphopoietin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033035.4 103 UTR 5 NP_149024.1
NM_138551.4 103 Intron NP_612561.2
XM_011543698.1 103 Intron XP_011542000.1

View Full Product Details