Product Details

SNP ID
rs142133240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:72443930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCGGCACAGTCTCTCTCCTGGCC[A/G]CCCTCTGCCCCAAGGTCACCCTTGC
Phenotype
MIM: 610159
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF366 PubMed Links

Gene Details

Gene
ZNF366
Gene Name
zinc finger protein 366
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152625.2 2247 Silent Mutation NP_689838.1

View Full Product Details