Product Details

SNP ID
rs142798897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:6372544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCCCCCCGGATGCTTCGATACT[C/T]AGCCATGTCTTCCGGAAATACTTTA
Phenotype
MIM: 612382
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MED10 PubMed Links

Gene Details

Gene
MED10
Gene Name
mediator complex subunit 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032286.2 410 Missense Mutation AAG,GAG K123E NP_115662.2

View Full Product Details