Product Details

SNP ID
rs143251724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:126870720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAAGGTACATTGACAGACTTTGG[A/G]ATGAGGAGAATCACCCTCTGATTGG
Phenotype
MIM: 613333
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MARCH3 PubMed Links

Gene Details

Gene
MARCH3
Gene Name
membrane associated ring-CH-type finger 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178450.4 1189 Silent Mutation ATC,ATT I225I NP_848545.1
XM_011543128.2 1189 Silent Mutation ATC,ATT I277I XP_011541430.1
XM_011543129.1 1189 Silent Mutation ATC,ATT I252I XP_011541431.1
XM_011543130.1 1189 Silent Mutation ATC,ATT I252I XP_011541432.1
XM_011543131.2 1189 Silent Mutation ATC,ATT I252I XP_011541433.1
XM_011543132.1 1189 Intron XP_011541434.1
XM_017009014.1 1189 Silent Mutation ATC,ATT I171I XP_016864503.1
XM_017009015.1 1189 Intron XP_016864504.1

View Full Product Details