Product Details

SNP ID
rs143676737
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:72444029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTTCCTCCAGCACCTCGGGGGC[A/G]TGCTCCGACTTGGTGGACAGATCCT
Phenotype
MIM: 610159
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ZNF366 PubMed Links

Gene Details

Gene
ZNF366
Gene Name
zinc finger protein 366
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152625.2 2148 Silent Mutation CAC,CAT H654H NP_689838.1

View Full Product Details