Product Details

SNP ID
rs146016179
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:152392334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCGGTCAGCTCCACAAAGTGGC[A/T]TTCTGTCAGGAAGATGTTCCGCTGG
Phenotype
MIM: 605108
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NMUR2 PubMed Links

Gene Details

Gene
NMUR2
Gene Name
neuromedin U receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020167.4 1271 Missense Mutation AGC,TGC S369C NP_064552.3
XM_011537670.2 1271 Intron XP_011535972.1

View Full Product Details