Product Details

SNP ID
rs146108114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:40681459 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGCTCTTTTGCGCGCTGCCCAAC[A/C]TGGGTCTCGGTAGCTCGCGGCTGCA
Phenotype
MIM: 601586
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PTGER4 PubMed Links

Gene Details

Gene
PTGER4
Gene Name
prostaglandin E receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000958.2 1058 Missense Mutation ATG,CTG M156L NP_000949.1
XM_017009656.1 1058 Missense Mutation ATG,CTG M156L XP_016865145.1
XM_017009657.1 1058 Missense Mutation ATG,CTG M156L XP_016865146.1
XM_017009658.1 1058 Missense Mutation ATG,CTG M156L XP_016865147.1
XM_017009659.1 1058 Missense Mutation ATG,CTG M156L XP_016865148.1

View Full Product Details