Product Details

SNP ID
rs147278612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:1802353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATAAAGACTACAGGAGAATTCG[G/T]TTTGTAGGTCGTCAGAAAGAGGTGA
Phenotype
MIM: 611842 MIM: 603848
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MRPL36 PubMed Links

Gene Details

Gene
MRPL36
Gene Name
mitochondrial ribosomal protein L36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032479.3 201 Intron NP_115868.1
XM_011514079.2 201 Intron XP_011512381.1
XM_011514080.2 201 Intron XP_011512382.1
XM_017009750.1 201 Intron XP_016865239.1
XM_017009751.1 201 Intron XP_016865240.1
XM_017009752.1 201 Intron XP_016865241.1
Gene
NDUFS6
Gene Name
NADH:ubiquinone oxidoreductase subunit S6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004553.4 201 Silent Mutation CGG,CGT R55R NP_004544.1

View Full Product Details