Product Details
- SNP ID
-
rs147511920
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:151259811 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGTGATGAAGGGAAGGACCCTGC[A/G]GTGATCAGAAGCCTGACTCTGGAGC
- Phenotype
-
MIM: 613109
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GM2A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs153477] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GM2A
- Gene Name
- GM2 ganglioside activator
View Full Product Details