Product Details

SNP ID
rs149329131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:54979338 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGAAATCACAGCCGGGATCAGCG[G/T]GGATTTAACCATTTCCTCATTACGG
Phenotype
MIM: 601521
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ESM1 PubMed Links

Gene Details

Gene
ESM1
Gene Name
endothelial cell specific molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135604.1 618 Silent Mutation CCA,CCC P133P NP_001129076.1
NM_007036.4 618 Silent Mutation CCA,CCC P183P NP_008967.1

View Full Product Details