Product Details

SNP ID
rs149510527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:9629470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCAAAACAGCAAAAAGGAAGATA[A/C]GCAATGGCACTGAGAACTCAGCAAC
Phenotype
MIM: 604796
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TAS2R1 PubMed Links

Gene Details

Gene
TAS2R1
Gene Name
taste 2 receptor member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019599.2 882 Missense Mutation CGT,CTT R188L NP_062545.1

View Full Product Details