Product Details

SNP ID
rs149936360
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140795377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCAATATTTCCAATGACAGTAA[A/G]GACTATCCGGTTTCCCGAATCAAGG
Phenotype
MIM: 606307 MIM: 606308 MIM: 606309
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCDHA1 PubMed Links

Gene Details

Gene
PCDHA1
Gene Name
protocadherin alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018900.3 519 Intron NP_061723.1
NM_031410.2 519 Intron NP_113598.1
NM_031411.2 519 Intron NP_113599.1
Gene
PCDHA2
Gene Name
protocadherin alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018905.2 519 Missense Mutation AAG,AGG K138R NP_061728.1
NM_031495.1 519 Missense Mutation AAG,AGG K138R NP_113683.1
NM_031496.1 519 Missense Mutation AAG,AGG K138R NP_113684.1
Gene
PCDHA3
Gene Name
protocadherin alpha 3
There are no transcripts associated with this gene.

View Full Product Details