Product Details

SNP ID
rs138852824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:108757178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTATGTGCAGGACACCATTCTAA[C/T]GTATACCTAAAATCTTCCCATGTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00222 PubMed Links

Gene Details

Gene
LINC00222
Gene Name
long intergenic non-protein coding RNA 222
There are no transcripts associated with this gene.

View Full Product Details