Product Details

SNP ID
rs139302046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:170537252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTACCTGGTTGTCAAGCAGGGG[C/T]TGTAGCATGGCACTTGCTGAGCCTC
Phenotype
MIM: 602017
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMB1 PubMed Links

Gene Details

Gene
PSMB1
Gene Name
proteasome subunit beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002793.3 609 Silent Mutation CAA,CAG Q174Q NP_002784.1

View Full Product Details