Product Details

SNP ID
rs140488380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27451550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTTGTCACATTCATTGCACTTA[C/T]AGGGCTTCTCCCCAGTGTGAATTCG
Phenotype
MIM: 602277
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF184 PubMed Links

Gene Details

Gene
ZNF184
Gene Name
zinc finger protein 184
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318891.1 2294 Missense Mutation TAT,TGT Y670C NP_001305820.1
NM_001318892.1 2294 Missense Mutation TAT,TGT Y670C NP_001305821.1
NM_001318893.1 2294 Missense Mutation TAT,TGT Y594C NP_001305822.1
NM_007149.2 2294 Missense Mutation TAT,TGT Y670C NP_009080.2
XM_011514861.2 2294 Missense Mutation TAT,TGT Y594C XP_011513163.1

View Full Product Details