Product Details

SNP ID
rs141063076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31954125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACTCCATCTTTTCATAGGTGAC[A/G]AAGGCACAGCTGGGATAAGAGAAAA
Phenotype
MIM: 613927 MIM: 138470 MIM: 154040 MIM: 600478
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2 PubMed Links

Gene Details

Gene
C2
Gene Name
complement component 2
There are no transcripts associated with this gene.

Gene
CFB
Gene Name
complement factor B
There are no transcripts associated with this gene.

Gene
MIR1236
Gene Name
microRNA 1236
There are no transcripts associated with this gene.

Gene
NELFE
Gene Name
negative elongation factor complex member E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002904.5 1101 Silent Mutation TTC,TTT F299F NP_002895.3
XM_006715205.3 1101 Silent Mutation TTC,TTT F294F XP_006715268.1
XM_011514913.2 1101 Silent Mutation TTC,TTT F244F XP_011513215.1
XM_017011299.1 1101 Silent Mutation TTC,TTT F239F XP_016866788.1
Gene
SKIV2L
Gene Name
Ski2 like RNA helicase
There are no transcripts associated with this gene.

View Full Product Details