Product Details

SNP ID
rs142527860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:110392924 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTACAGGCAGCCTCTGTCCATCTC[C/G]CGCAAGGAGCTCTGTCTGCAGTCGC
Phenotype
MIM: 124450
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DDO PubMed Links

Gene Details

Gene
DDO
Gene Name
D-aspartate oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003649.2 788 Missense Mutation CGA,GGA R321G NP_003640.2
NM_004032.2 788 Missense Mutation CGA,GGA R262G NP_004023.2

View Full Product Details