Product Details

SNP ID
rs142807196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30137205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGTCTTCTGCTGCCCCCTCTGC[C/T]GGAAGCCCTGTTCTGAGGAGGTGCT
Phenotype
MIM: 616976
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIM40 PubMed Links

Gene Details

Gene
TRIM40
Gene Name
tripartite motif containing 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286633.1 986 Missense Mutation CGG,TGG R57W NP_001273562.1
NM_138700.4 986 Missense Mutation CGG,TGG R57W NP_619645.1
XM_011514305.1 986 Missense Mutation CGG,TGG R57W XP_011512607.1
XM_011514306.1 986 Missense Mutation CGG,TGG R57W XP_011512608.1
XM_011514308.1 986 Missense Mutation CGG,TGG R57W XP_011512610.1
XM_011514309.1 986 Missense Mutation CGG,TGG R57W XP_011512611.1

View Full Product Details