Product Details

SNP ID
rs142977156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:132459615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGAGCTTGCCAAGTTAAACACC[A/G]CCTTCAGTACCGAGCTGCTATACAA
Phenotype
MIM: 603217
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STX7 PubMed Links

Gene Details

Gene
STX7
Gene Name
syntaxin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001326578.1 2179 UTR 3 NP_001313507.1
NM_001326579.1 2179 UTR 3 NP_001313508.1
NM_001326580.1 2179 Intron NP_001313509.1
NM_003569.2 2179 UTR 3 NP_003560.2
XM_011536178.1 2179 Intron XP_011534480.1

View Full Product Details