Product Details

SNP ID
rs143342827
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:159977258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTTGAGTTTGTCACCATGATTG[C/T]ATGGTGCTGGTGCCCTTGCAGTGAC
Phenotype
MIM: 147280
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
IGF2R PubMed Links

Gene Details

Gene
IGF2R
Gene Name
insulin like growth factor 2 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000876.3 Intron NP_000867.2

View Full Product Details