Product Details

SNP ID
rs144275406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:159780007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTTCACTACACAAAGTGCATCA[C/T]GTAAAGAGCGCTCCATCTCATCACA
Phenotype
MIM: 100678 MIM: 611831 MIM: 186980
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACAT2 PubMed Links

Gene Details

Gene
ACAT2
Gene Name
acetyl-CoA acetyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303253.1 1413 Intron NP_001290182.1
NM_005891.2 1413 Intron NP_005882.2
Gene
MRPL18
Gene Name
mitochondrial ribosomal protein L18
There are no transcripts associated with this gene.

Gene
SNORA20
Gene Name
small nucleolar RNA, H/ACA box 20
There are no transcripts associated with this gene.

Gene
SNORA29
Gene Name
small nucleolar RNA, H/ACA box 29
There are no transcripts associated with this gene.

Gene
TCP1
Gene Name
t-complex 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008897.1 1413 Missense Mutation CAT,CGT H238R NP_001008897.1
NM_030752.2 1413 Missense Mutation CAT,CGT H393R NP_110379.2

View Full Product Details