Product Details

SNP ID
rs144434700
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:167157198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCGCTGAAGCTTGGGCTGTTTCT[C/T]AGGCTCCCGGAGTCTTTTCTGGAGA
Phenotype
MIM: 602043
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR31 PubMed Links

Gene Details

Gene
GPR31
Gene Name
G protein-coupled receptor 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005299.2 634 Missense Mutation AAG,GAG K212E NP_005290.2
Gene
LOC105378123
Gene Name
uncharacterized LOC105378123
There are no transcripts associated with this gene.

View Full Product Details