Product Details

SNP ID
rs145222586
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116647517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACACACTACCTTTGGACTCCCTTC[A/T]CCCTCTGAAGAATAATAGTTCAATA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZUFSP PubMed Links

Gene Details

Gene
ZUFSP
Gene Name
zinc finger with UFM1 specific peptidase domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145062.2 2063 Silent Mutation GGA,GGT G470G NP_659499.2
XM_011535565.2 2063 Silent Mutation GGA,GGT G470G XP_011533867.1
XM_011535567.1 2063 Silent Mutation GGA,GGT G302G XP_011533869.1
XM_017010393.1 2063 Silent Mutation GGA,GGT G470G XP_016865882.1
XM_017010394.1 2063 Silent Mutation GGA,GGT G274G XP_016865883.1
XM_017010395.1 2063 Silent Mutation GGA,GGT G246G XP_016865884.1
XM_017010396.1 2063 Silent Mutation GGA,GGT G246G XP_016865885.1

View Full Product Details