Product Details

SNP ID
rs145569223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:83216565 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGGATAAAGCCGACCCTCTTCC[A/C]AGTGTTTATCTGACACTTGCTGAGC
Phenotype
MIM: 154250
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ME1 PubMed Links

Gene Details

Gene
ME1
Gene Name
malic enzyme 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002395.5 1746 Missense Mutation TGG,TTG W494L NP_002386.1
XM_011535836.2 1746 Missense Mutation TGG,TTG W419L XP_011534138.1

View Full Product Details