Product Details

SNP ID
rs146154106
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:24544372 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACACCTTTATTTAGATTAGTGCAT[A/G]TATTTAATAGTGTGATATTAGAAAT
Phenotype
MIM: 610045 MIM: 609269
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH5A1 PubMed Links

Gene Details

Gene
ALDH5A1
Gene Name
aldehyde dehydrogenase 5 family member A1
There are no transcripts associated with this gene.

Gene
KIAA0319
Gene Name
KIAA0319
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168374.1 5047 UTR 3 NP_001161846.1
NM_001168375.1 5047 UTR 3 NP_001161847.1
NM_001168376.1 5047 UTR 3 NP_001161848.1
NM_001168377.1 5047 UTR 3 NP_001161849.1
NM_001252328.1 5047 UTR 3 NP_001239257.1
NM_014809.3 5047 UTR 3 NP_055624.2
XM_011515025.2 5047 UTR 3 XP_011513327.1
XM_011515026.2 5047 UTR 3 XP_011513328.1
XM_017011541.1 5047 UTR 3 XP_016867030.1
XM_017011542.1 5047 Intron XP_016867031.1
XM_017011543.1 5047 UTR 3 XP_016867032.1
XM_017011544.1 5047 UTR 3 XP_016867033.1
XM_017011545.1 5047 UTR 3 XP_016867034.1
XM_017011546.1 5047 Intron XP_016867035.1
XM_017011547.1 5047 UTR 3 XP_016867036.1
XM_017011548.1 5047 UTR 3 XP_016867037.1
XM_017011549.1 5047 UTR 3 XP_016867038.1
XM_017011550.1 5047 Intron XP_016867039.1
XM_017011551.1 5047 UTR 3 XP_016867040.1
XM_017011552.1 5047 UTR 3 XP_016867041.1
XM_017011553.1 5047 UTR 3 XP_016867042.1

View Full Product Details