Product Details

SNP ID
rs146214653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33076092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCCGGACAGTGGCTCTGACGGC[A/G]TTACTGATGGTGCTGCTCACATCTG
Phenotype
MIM: 142880 MIM: 142858
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HLA-DPA1 PubMed Links

Gene Details

Gene
HLA-DPA1
Gene Name
major histocompatibility complex, class II, DP alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242524.1 167 Intron NP_001229453.1
NM_001242525.1 167 Intron NP_001229454.1
NM_033554.3 167 Intron NP_291032.2
Gene
HLA-DPB1
Gene Name
major histocompatibility complex, class II, DP beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002121.5 167 Silent Mutation GCA,GCG A17A NP_002112.3

View Full Product Details