Product Details

SNP ID
rs146220191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:7883178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTTCGCTTGGCTCAGGACAAAG[C/T]GGTGTAACGAGTCAAGGTCTCTGCC
Phenotype
MIM: 112266 MIM: 616412
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BLOC1S5-TXNDC5 PubMed Links

Gene Details

Gene
BLOC1S5-TXNDC5
Gene Name
BLOC1S5-TXNDC5 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
BMP6
Gene Name
bone morphogenetic protein 6
There are no transcripts associated with this gene.

Gene
TXNDC5
Gene Name
thioredoxin domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145549.2 1303 Missense Mutation CAC,CGC H314R NP_001139021.1
NM_030810.3 1303 Missense Mutation CAC,CGC H422R NP_110437.2

View Full Product Details