Product Details

SNP ID
rs146316351
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:40391989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTCGGAGCTGCCAAAAGTCCCC[C/T]GGGCCCCCACCAGGTCACTCTCCTC
Phenotype
MIM: 612808
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRFN2 PubMed Links

Gene Details

Gene
LRFN2
Gene Name
leucine rich repeat and fibronectin type III domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020737.2 3735 Missense Mutation NP_065788.1
XM_011514761.2 3735 Missense Mutation XP_011513063.1
XM_011514762.2 3735 Missense Mutation XP_011513064.1
XM_017011110.1 3735 Missense Mutation XP_016866599.1
XM_017011111.1 3735 Intron XP_016866600.1

View Full Product Details