Product Details

SNP ID
rs147062235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29306914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACCTCTGCTTCATCTCTGTCAC[A/G]GTCCCCCAGTCCATTGCAAATTCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR14J1 PubMed Links

Gene Details

Gene
OR14J1
Gene Name
olfactory receptor family 14 subfamily J member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030946.1 225 Silent Mutation ACA,ACG T75T NP_112208.1

View Full Product Details