Product Details
- SNP ID
-
rs147083506
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:149400435 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAGAAGAAGTCAAGACTGAGAACA[A/G]CAATCATATTAATTTGAAGGTGGCG
- Phenotype
-
MIM: 608829
MIM: 605101
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SUMO4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs237025] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SUMO4
- Gene Name
- small ubiquitin-like modifier 4
- Gene
- TAB2
- Gene Name
- TGF-beta activated kinase 1/MAP3K7 binding protein 2
View Full Product Details