Product Details

SNP ID
rs147307438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26184143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGCACGCTGTGTCAGAGGGCAC[C/T]AAGGCCGTTACCAAGTACACCAGCT
Phenotype
MIM: 602805 MIM: 602823
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HIST1H2BE PubMed Links

Gene Details

Gene
HIST1H2BE
Gene Name
histone cluster 1, H2be
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003523.2 Intron NP_003514.2
Gene
HIST1H4D
Gene Name
histone cluster 1, H4d
There are no transcripts associated with this gene.

View Full Product Details