Product Details

SNP ID
rs147622332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32937287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGGTCTGGTATGTCCAGTCTCC[A/G]TTGGGCTGGGCAGTCTTGTGCGCAC
Phenotype
MIM: 142856
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HLA-DMB PubMed Links

Gene Details

Gene
HLA-DMB
Gene Name
major histocompatibility complex, class II, DM beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002118.4 770 Silent Mutation AAC,AAT N169N NP_002109.2

View Full Product Details