Product Details
- SNP ID
-
rs147891075
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:26031719 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGCATAATAGTCACTCGCTTAGC[A/G]TGGATGGCGCAAAGGTTTGTGTCCT
- Phenotype
-
MIM: 602795
MIM: 602819
MIM: 602822
MIM: 602829
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HIST1H2AB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2213284,rs34966100] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HIST1H2AB
- Gene Name
- histone cluster 1, H2ab
There are no transcripts associated with this gene.
- Gene
- HIST1H3B
- Gene Name
- histone cluster 1, H3b
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003537.3 |
|
Intron |
|
|
NP_003528.1 |
- Gene
- HIST1H4A
- Gene Name
- histone cluster 1, H4a
There are no transcripts associated with this gene.
- Gene
- HIST1H4B
- Gene Name
- histone cluster 1, H4b
There are no transcripts associated with this gene.
View Full Product Details