Product Details

SNP ID
rs147896204
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:82365513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAACTGATCCTGAACCACATCGT[C/G]CCCCCTGAAGATGAGCGGCAGAACC
Phenotype
MIM: 190920
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TPBG PubMed Links

Gene Details

Gene
TPBG
Gene Name
trophoblast glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166392.1 977 Silent Mutation GTC,GTG V184V NP_001159864.1
NM_006670.4 977 Silent Mutation GTC,GTG V184V NP_006661.1
XM_011536097.2 977 Silent Mutation GTC,GTG V184V XP_011534399.1

View Full Product Details